Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecafluorooctanenitrile 96.0+%, TCI America™
SDP

Catalog No. P24831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P24831G Supplier TCI America Supplier No. P24831G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 647-12-1
Color Yellow
Boiling Point 102°C
Molecular Formula C8F15N
Quantity 1 g
MDL Number MFCD00039481
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluorooctanenitrile, Perfluorooctanonitrile
InChI Key FULLNJNBFUCVES-UHFFFAOYSA-N
SMILES C(#N)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanenitrile
Molecular Weight (g/mol) 395.071
PubChem CID 69536
Formula Weight 395.07
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentadecafluorooctanenitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.