Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentadecafluorooctanoic acid ammonium salt

Catalog No. 7726250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-262-50G 50 g
77-262-10G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-262-50G Supplier Sigma Aldrich Supplier No. 7726250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentadecafluorooctanoic acid ammonium salt

Specifications

CAS 3825-26-1
Molecular Formula C8H4F15NO2
Linear Formula CF3(CF2)6 COONH4
MDL Number MFCD00042599
Quantity 50 g
Synonym Ammonium pentadecafluorooctanoate; Perfluorocaprylic acid ammonium salt; Perfluorooctanoic acid ammonium salt
Molecular Weight (g/mol) 431.1
Percent Purity ≥98.0% (NT)
Biological Activity Liver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.