Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentadecafluorooctanoyl Chloride

Catalog No. 503081445 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1445 5 g
18-989-8100ML 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 50-308-1445 Supplier Sigma Aldrich Supplier No. 2909045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentadecafluorooctanoyl chloride can be used:In the derivatization of poly(2-hydroxyethyl methacrylate) (PHEMA) via esterification for use as composite membranes.In the fabrication of superhydrophobic cellulose surfaces.As a reagent for the esterification of hydroxyl-functionalized gold nanocrystals.As a reagent in the synthesis of fluorous derivatives of diaminocyclohexane which are used as ligands.

Specifications

CAS 335-64-8
Density 1.744 g/mL at 25°C
Boiling Point 129°C to 130°C (744 mmHg, lit.)
Molecular Formula C8ClF15O
Refractive Index n20/D 1.3045
Linear Formula CF3(CF2)6COCl
MDL Number MFCD00013654
Quantity 5 g
Molecular Weight (g/mol) 432.51 g/mol
Biological Activity Liver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.