Learn More
Medchemexpress LLC ω-pentadecalactone | 106-02-5 | MFCD00039667 | 240.38 g/mol | C15H28O2 | 25 G
SDP
Catalog No. 5000302851
Encompass_Preferred
ω-Pentadecalactone is a macrocyclic lactone fragrance ingredient used in research and formulation. It is a 15-membered lactone (C15H28O2) with a characteristic musk-like, vanilla-type odor and documented physical properties for analytical and formulation work.
  • Macrocyclic lactone structure with musk-like, vanilla-type odor.
  • Used as a fragrance ingredient and research reagent.
  • Defined physical properties: melting point ~32 °C and boiling point 137 °C at 2 mmHg.
  • Stable under recommended storage conditions (-20 °C to 4 °C for extended storage).
  • GHS aquatic toxicity classifications H400 and H411; avoid release to the environment.
Catalog No. 50-003-02851 Supplier Medchemexpress LLC Supplier No. 25GHYW035362
ω-Pentadecalactone is a macrocyclic lactone fragrance ingredient used in research and formulation. It is a 15-membered lactone (C15H28O2) with a characteristic musk-like, vanilla-type odor and documented physical properties for analytical and formulation work.
  • Macrocyclic lactone structure with musk-like, vanilla-type odor.
  • Used as a fragrance ingredient and research reagent.
  • Defined physical properties: melting point ~32 °C and boiling point 137 °C at 2 mmHg.
  • Stable under recommended storage conditions (-20 °C to 4 °C for extended storage).
  • GHS aquatic toxicity classifications H400 and H411; avoid release to the environment.