Learn More
Medchemexpress LLC ω-pentadecalactone | 106-02-5 | MFCD00039667 | 99.7% | 240.38 g/mol | C15H28O2 | 100 G
SDP
Catalog No. 5000299401
Encompass_Preferred
ω-Pentadecalactone is a macrocyclic lactone used as a fragrance ingredient and supplied as a high-purity analytical standard for research use. It has chemical formula C15H28O2, molecular weight 240.38 g/mol, and a reported purity of about 99.71%.
  • Macrocyclic lactone with fragrance applications in perfumery and flavor research.
  • High reported purity (≈99.7%), suitable for analytical and reference use.
  • Solid material with defined storage recommendations to preserve stability.
  • Soluble in common organic solvents such as ethanol and DMSO.
  • Provided with a safety data sheet and handling instructions for laboratory use.
Catalog No. 50-002-99401 Supplier Medchemexpress LLC Supplier No. HYW035362100G
ω-Pentadecalactone is a macrocyclic lactone used as a fragrance ingredient and supplied as a high-purity analytical standard for research use. It has chemical formula C15H28O2, molecular weight 240.38 g/mol, and a reported purity of about 99.71%.
  • Macrocyclic lactone with fragrance applications in perfumery and flavor research.
  • High reported purity (≈99.7%), suitable for analytical and reference use.
  • Solid material with defined storage recommendations to preserve stability.
  • Soluble in common organic solvents such as ethanol and DMSO.
  • Provided with a safety data sheet and handling instructions for laboratory use.