Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentadecane | 629-62-9 | 50 G
SDP

Catalog No. 5000419655
Encompass_Preferred

Pentadecane is a floral volatile antimalarial agent that inhibits the growth of parasites such as Leishmania and prevents macrophage infection. Its IC50s against promastigotes and amastigotes are 65.3 μM and 60.5 μM, respectively. Pentadecane also possesses anti-inflammatory, analgesic, and antipyretic activities.

  • Inhibits growth of parasites such as Leishmania
  • Prevents macrophage infection
  • Exhibits anti-inflammatory activities
  • Exhibits analgesic activities
  • Exhibits antipyretic activities
  • Appearance: Liquid, colorless to off-white
  • Purity: 98.88%

Catalog No. 50-004-19655 Supplier Medchemexpress LLC Supplier No. HYN792650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentadecane is a floral volatile antimalarial agent that inhibits the growth of parasites such as Leishmania and prevents macrophage infection. Its IC50s against promastigotes and amastigotes are 65.3 μM and 60.5 μM, respectively. Pentadecane also possesses anti-inflammatory, analgesic, and antipyretic activities.

  • Inhibits growth of parasites such as Leishmania
  • Prevents macrophage infection
  • Exhibits anti-inflammatory activities
  • Exhibits analgesic activities
  • Exhibits antipyretic activities
  • Appearance: Liquid, colorless to off-white
  • Purity: 98.88%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.