Learn More
Medchemexpress LLC Pentadecane | 629-62-9 | MFCD00008990 | 100 G
SDP
Catalog No. 5000417745
Encompass_Preferred
Pentadecane is a floral volatile antimalarial agent that can inhibit the growth of parasites such as Leishmania and avoid infection of macrophages. It has IC50s against promastigotes and amastigotes of 65.3 μM and 60.5 μM, respectively. This agent also exhibits anti-inflammatory, analgesic, and antipyretic activities.
  • It is a floral volatile antimalarial agent.
  • It inhibits the growth of parasites such as Leishmania.
  • It helps avoid infection of macrophages.
  • It possesses anti-inflammatory activities.
  • It has analgesic properties.
  • It demonstrates antipyretic activities.
Storage Conditions: Store at room temperature for 3 years. When in solvent, store at -80°C for 2 years or at -20°C for 1 year. For stock solutions, store at -80°C for 2 years or at -20°C for 1 year.
Catalog No. 50-004-17745 Supplier Medchemexpress LLC Supplier No. HYN7926100G
Pentadecane is a floral volatile antimalarial agent that can inhibit the growth of parasites such as Leishmania and avoid infection of macrophages. It has IC50s against promastigotes and amastigotes of 65.3 μM and 60.5 μM, respectively. This agent also exhibits anti-inflammatory, analgesic, and antipyretic activities.
  • It is a floral volatile antimalarial agent.
  • It inhibits the growth of parasites such as Leishmania.
  • It helps avoid infection of macrophages.
  • It possesses anti-inflammatory activities.
  • It has analgesic properties.
  • It demonstrates antipyretic activities.
Storage Conditions: Store at room temperature for 3 years. When in solvent, store at -80°C for 2 years or at -20°C for 1 year. For stock solutions, store at -80°C for 2 years or at -20°C for 1 year.