Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentadecanedioic acid, 1,15-dimethyl ester | 36575-82-3 | 99.0% | 300.43 g/mol | C17H32O4 | 100 G
SDP

Catalog No. 5000280964
Encompass_Preferred

Dimethyl pentadecanedioate (pentadecanedioic acid, 1,15-dimethyl ester) is the dimethyl ester derivative of pentadecanedioic acid supplied for laboratory research. It is used as a reagent and synthetic intermediate in organic synthesis and biochemical studies. The material is provided with quality documentation and is intended for research use only, not for human or clinical use.

  • High purity (≈99.0%).
  • CAS number 36575-82-3 for unambiguous identification.
  • Molecular weight 300.43 g/mol.
  • Chemical formula C17H32O4.
  • Supplied with certificate of analysis for batch verification.
  • Available in gram-scale packaging for laboratory use.
  • Suitable as a synthetic intermediate and research reagent.

Catalog No. 50-002-80964 Supplier Medchemexpress LLC Supplier No. HYW460119100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl pentadecanedioate (pentadecanedioic acid, 1,15-dimethyl ester) is the dimethyl ester derivative of pentadecanedioic acid supplied for laboratory research. It is used as a reagent and synthetic intermediate in organic synthesis and biochemical studies. The material is provided with quality documentation and is intended for research use only, not for human or clinical use.

  • High purity (≈99.0%).
  • CAS number 36575-82-3 for unambiguous identification.
  • Molecular weight 300.43 g/mol.
  • Chemical formula C17H32O4.
  • Supplied with certificate of analysis for batch verification.
  • Available in gram-scale packaging for laboratory use.
  • Suitable as a synthetic intermediate and research reagent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.