Learn More
Medchemexpress LLC Pentadecanedioic acid, 1,15-dimethyl ester | 36575-82-3 | MFCD00673462 | 99.0% | 300.43 g/mol | C17H32O4 | 25 G
SDP
Catalog No. 5000281402
Encompass_Preferred
Dimethyl pentadecanedioate is the dimethyl ester of pentadecanedioic acid used as a research reagent and synthetic intermediate. It is supplied as a solid with CAS number 36575-82-3 and a GC-reported purity of 98.95% (COA). Typical applications include use as a building block in organic synthesis and materials research.
  • High purity suitable for research applications (GC purity 98.9%).
  • Dimethyl ester functionality useful as a synthetic intermediate.
  • Solid form with a melting point near 43-44 °C for easy handling.
  • Soluble in common organic solvents such as ethanol and chloroform.
  • Available in laboratory pack sizes including 25 G.
Catalog No. 50-002-81402 Supplier Medchemexpress LLC Supplier No. HYW46011925G
Dimethyl pentadecanedioate is the dimethyl ester of pentadecanedioic acid used as a research reagent and synthetic intermediate. It is supplied as a solid with CAS number 36575-82-3 and a GC-reported purity of 98.95% (COA). Typical applications include use as a building block in organic synthesis and materials research.
  • High purity suitable for research applications (GC purity 98.9%).
  • Dimethyl ester functionality useful as a synthetic intermediate.
  • Solid form with a melting point near 43-44 °C for easy handling.
  • Soluble in common organic solvents such as ethanol and chloroform.
  • Available in laboratory pack sizes including 25 G.