Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecanoic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012271 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2271 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2271 Supplier MilliporeSigma Supelco Supplier No. 914465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentadecanoic acid is a saturated fatty acid, commonly present in ox bile. It is present as a phytochemical component of Indigofera suffruticosa leaves and is known to exhibit antimicrobial and antioxidant properties.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1002-84-2
Flash Point >113°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)13 COOH
MDL Number MFCD00002745
Melting Point 51°C to 53°C; 51°C to 55°C
Boiling Point 257°C (100 mmHg)
Synonym C15:0; NSC 28486; Pentadecylic acid
Type Analytical Standard
Quantity 5 g
Formula Weight 242.40
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.