Learn More
Medchemexpress LLC Pentadecanoic acid-d29 | 130348-95-7 | MFCD00053332 | 271.58 | 10 MG
SDP
Catalog No. 5000447512
Encompass_Preferred
Pentadecanoic acid-d29 is the deuterium labeled Pentadecanoic acid. Pentadecylic acid is a saturated fatty acid with a 15-carbon backbone. It is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-47512 Supplier Medchemexpress LLC Supplier No. HYW004283S10MG
Pentadecanoic acid-d29 is the deuterium labeled Pentadecanoic acid. Pentadecylic acid is a saturated fatty acid with a 15-carbon backbone. It is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS