Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentadecanoic-d29 acid, 1,2,3-propanetriyl ester | 352431-43-7 | 99.2% | C48H5D87O6 | 10MG
SDP

Catalog No. 5000258629
Encompass_Preferred

Tripentadecanoin-d29 is a deuterium-labeled triglyceride standard used as an isotopic tracer and internal standard for quantitative lipid analysis. It provides mass-shifted fatty acyl chains for unambiguous detection and accurate quantification in mass spectrometry and NMR workflows.

  • Deuterium-labeled triglyceride standard for analytical and tracer applications.
  • Suitable as an internal standard for quantitative mass spectrometry and NMR.
  • High isotopic enrichment (d29) for clear mass distinction from natural lipids.
  • High purity (99.2%) for reliable quantification and reproducibility.
  • Supplied in small analytical quantity (10 mg) for method development and validation.
  • Molecular formula C48H5D87O6, molecular weight 852.78.

Catalog No. 50-002-58629 Supplier Medchemexpress LLC Supplier No. HY146726S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tripentadecanoin-d29 is a deuterium-labeled triglyceride standard used as an isotopic tracer and internal standard for quantitative lipid analysis. It provides mass-shifted fatty acyl chains for unambiguous detection and accurate quantification in mass spectrometry and NMR workflows.

  • Deuterium-labeled triglyceride standard for analytical and tracer applications.
  • Suitable as an internal standard for quantitative mass spectrometry and NMR.
  • High isotopic enrichment (d29) for clear mass distinction from natural lipids.
  • High purity (99.2%) for reliable quantification and reproducibility.
  • Supplied in small analytical quantity (10 mg) for method development and validation.
  • Molecular formula C48H5D87O6, molecular weight 852.78.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.