Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecanonitrile 98.0+%, TCI America™
SDP

Catalog No. P087125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P087125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P087125G Supplier TCI America Supplier No. P087125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18300-91-9
Molecular Formula C15H29N
Molecular Weight (g/mol) 223.40
MDL Number MFCD00019849
InChI Key KRKQHNVYOWTEQO-UHFFFAOYSA-N
Synonym Myristyl Cyanide, Tetradecyl Cyanide
PubChem CID 87568
IUPAC Name pentadecanenitrile
SMILES CCCCCCCCCCCCCCC#N

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 223.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Pentadecanonitrile
EINECSNumber (2)-1507
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.