Learn More
Cayman Chemical PentadecnoIc AcId-d29 1mg
SDP
Supplier:  Cayman Chemical 280811 MG
Encompass_Preferred
An internal standard for the quantification of pentadecanoic acid by GC- or LC-MS
Catalog No. 50-293-7628