Learn More
Cayman Chemical PentadecnoIc AcId-d3 10mg
SDP
Catalog No. 502937061
Encompass_Preferred
An internal standard for the quantification of pentadecanoic acid by GC- or LC-MS
Catalog No. 50-293-7061 Supplier Cayman Chemical Supplier No. 2859610 MG
Add to Cart
Add to Cart
An internal standard for the quantification of pentadecanoic acid by GC- or LC-MS