Learn More
Cayman Chemical Pentadecanoic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 352431-40-4, 1 mg
SDP
Catalog No. 502938517
Encompass_Preferred
An internal standard for the quantification of pentadecanoic acid by GC- or LC-MS
Catalog No. 50-293-8517 Supplier Cayman Chemical Supplier No. 285961 MG
An internal standard for the quantification of pentadecanoic acid by GC- or LC-MS