Learn More
Cayman Chemical Pentadecanoic Acid methyl ester, Purity: ≥95%, CAS: 7132-64-1, 1 g
SDP
Catalog No. 502935059
Encompass_Preferred
An esterified form of pentadecanoic acid; has been found as a minor component in biodiesels made from the transesterification of beef tallow, soybean oil, and babassu oil blends; has been found in latent fingerprint residue samples
Catalog No. 50-293-5059 Supplier Cayman Chemical Supplier No. 267221 G
An esterified form of pentadecanoic acid; has been found as a minor component in biodiesels made from the transesterification of beef tallow, soybean oil, and babassu oil blends; has been found in latent fingerprint residue samples