Learn More
Cayman Chemical Pentadecanoic Acid methyl ester, Purity: ≥95%, CAS: 7132-64-1, 500 mg
SDP
Catalog No. 502934191
Encompass_Preferred
An esterified form of pentadecanoic acid; has been found as a minor component in biodiesels made from the transesterification of beef tallow, soybean oil, and babassu oil blends; has been found in latent fingerprint residue samples
Catalog No. 50-293-4191 Supplier Cayman Chemical Supplier No. 26722500 MG
An esterified form of pentadecanoic acid; has been found as a minor component in biodiesels made from the transesterification of beef tallow, soybean oil, and babassu oil blends; has been found in latent fingerprint residue samples