Learn More
Cayman Chemical Pentadecanoyl Ethanolamide, Purity: ≥98%, CAS: 53832-58-9, 50 mg
SDP
Catalog No. 502893136
Encompass_Preferred
A member of the family of fatty N-acyl ethanolamines collectively called endocannabinoids
Catalog No. 50-289-3136 Supplier Cayman Chemical Supplier No. 900174050 MG
A member of the family of fatty N-acyl ethanolamines collectively called endocannabinoids