Learn More
Cayman Chemical Pentadecnoyl-L-carnItInch 10mg
SDP
Catalog No. 502916430
Encompass_Preferred
A CB receptor agonist (EC50s = 3.7 and 3.2 µM for CB1 and CB2 receptors, respectively, in a cAMP modulation assay); a 5-HT1A and 5-HT1B agonist (EC50s = 9.6 and 9.3 µM, respectively); a histamine H1 and H2 antagonist (EC50s = 10.9 and 6.1 µM, respectively); inhibits T cell proliferation in an SAg primary human cell system; reduces CCL2 levels in an Mphg primary human cell system
Catalog No. 50-291-6430 Supplier Cayman Chemical Supplier No. 3767710 MG
A CB receptor agonist (EC50s = 3.7 and 3.2 µM for CB1 and CB2 receptors, respectively, in a cAMP modulation assay); a 5-HT1A and 5-HT1B agonist (EC50s = 9.6 and 9.3 µM, respectively); a histamine H1 and H2 antagonist (EC50s = 10.9 and 6.1 µM, respectively); inhibits T cell proliferation in an SAg primary human cell system; reduces CCL2 levels in an Mphg primary human cell system