Learn More
Medchemexpress LLC Pentaerythritol dist 50g
SDP
Catalog No. 5000206605
Encompass_Preferred
Pentaerythritol distearate (mixture) is a mixture of ester compounds commonly used as a thickener emulsifier or lubricant in various cosmetic and personal care products It is also used as a plasticizer or stabilizer in the production of plastics and resins Pentaerythritol distearate (mixture) has unique chemical properties that make it an important ingredient in a variety of industrial processes including the manufacture of adhesives coatings and sealants
Catalog No. 50-002-06605 Supplier Medchemexpress LLC Supplier No. HYW12736450G
Pentaerythritol distearate (mixture) is a mixture of ester compounds commonly used as a thickener emulsifier or lubricant in various cosmetic and personal care products It is also used as a plasticizer or stabilizer in the production of plastics and resins Pentaerythritol distearate (mixture) has unique chemical properties that make it an important ingredient in a variety of industrial processes including the manufacture of adhesives coatings and sealants