Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaerythritol Distearate (so called) 95.0+%, TCI America™
SDP

Catalog No. P073825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P073825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P073825G Supplier TCI America Supplier No. P073825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13081-97-5
Molecular Formula C41H80O6
Molecular Weight (g/mol) 669.085
MDL Number MFCD00059225
InChI Key FSEJJKIPRNUIFL-UHFFFAOYSA-N
PubChem CID 61575
IUPAC Name [2,2-bis(hydroxymethyl)-3-octadecanoyloxypropyl] octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(CO)(CO)COC(=O)CCCCCCCCCCCCCCCCC

Specifications

Melting Point 72°C
Quantity 25 g
Formula Weight 669.09
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pentaerythritol Distearate (so called) [Lubricants]
EINECSNumber (2)-0642&(2)-0766
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.