Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentaerythritol propoxylate (5/4 PO/OH)

Catalog No. 418749500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-874-9500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-874-9500ML Supplier Sigma Aldrich Supplier No. 418749500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentaerythritol propoxylate (5/4 PO/OH) is a pentaerythriol based branched polymer that can be used as a crystallization agent to generate ordered, high resolution diffraction quality crystals.Pentaerythritol propoxylate may be used as a precipitant in reservoir solution for the crystallization of Phosphate Transporter(PiPT) and Aggregation Suppressing Protein A(AgsA). It may also acts as a reservoir solution for Phytosulfokine (PSK)- receptor PSKR- and somatic embryogenesis receptor-like kinases (SERKs) complex.

Specifications

CAS 9051-49-4
Density 1.05 g/mL (at 25°C)
Boiling Point >300°C
Refractive Index n20/D 1.464
Linear Formula C[CH2[OCH2CH(CH3)]nOH]4
MDL Number MFCD00197885
Quantity 500 mL
Synonym Pentaerythritol propoxylate (17/8 PO/OH)
Molecular Weight (g/mol) 426
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.