Learn More
Medchemexpress LLC Pentaerythritol tetra(3-mercaptopropionate) | 7575-23-7 | MFCD00022104 | 95.0% | 488.66 g/mol | C17H28O8S4 | 100 G
SDP
Catalog No. 5000281679
Encompass_Preferred
Pentaerythritol tetra(3-mercaptopropionate) is a tetrafunctional thiol reagent used as a crosslinking agent and multifunctional building block in polymer and materials chemistry. It enables rapid formation of network polymers through thiol-ene reactions and is commonly used in curing formulations and materials synthesis where controlled crosslink density is required.
  • Multifunctional tetra-thiol reagent for crosslinking and network formation.
  • Facilitates thiol-ene click chemistry for efficient polymerization.
  • Enables tuning of mechanical and thermal properties in cured materials.
  • Liquid form allows easy handling and incorporation into formulations.
  • Supplied with characterization documents to support reproducible use.
Catalog No. 50-002-81679 Supplier Medchemexpress LLC Supplier No. HYW250309100G
Pentaerythritol tetra(3-mercaptopropionate) is a tetrafunctional thiol reagent used as a crosslinking agent and multifunctional building block in polymer and materials chemistry. It enables rapid formation of network polymers through thiol-ene reactions and is commonly used in curing formulations and materials synthesis where controlled crosslink density is required.
  • Multifunctional tetra-thiol reagent for crosslinking and network formation.
  • Facilitates thiol-ene click chemistry for efficient polymerization.
  • Enables tuning of mechanical and thermal properties in cured materials.
  • Liquid form allows easy handling and incorporation into formulations.
  • Supplied with characterization documents to support reproducible use.