Learn More
Medchemexpress LLC Pentaerythritol tetra(3-mercaptopropionate) | 7575-23-7 | MFCD00022104 | 95.0% | 488.66 g/mol | C17H28O8S4 | 25 G
SDP
Catalog No. 5000281242
Encompass_Preferred
Pentaerythritol tetra(3-mercaptopropionate) is a tetra-functional thiol reagent used as a crosslinker and building block in polymer chemistry and materials research. It enables efficient thiol-ene curing and formation of dense, thiol-containing networks suitable for coatings, adhesives, and specialty polymer synthesis.
  • Tetra-functional thiol functionality for high crosslink density.
  • Facilitates thiol-ene and click chemistry curing.
  • Improves mechanical strength and thermal stability of polymer networks.
  • Liquid, colorless to light-yellow appearance.
  • Soluble in DMSO; may require sonication.
  • Typical purity around 95%.
  • Store refrigerated under inert atmosphere.
Catalog No. 50-002-81242 Supplier Medchemexpress LLC Supplier No. HYW25030925G
Pentaerythritol tetra(3-mercaptopropionate) is a tetra-functional thiol reagent used as a crosslinker and building block in polymer chemistry and materials research. It enables efficient thiol-ene curing and formation of dense, thiol-containing networks suitable for coatings, adhesives, and specialty polymer synthesis.
  • Tetra-functional thiol functionality for high crosslink density.
  • Facilitates thiol-ene and click chemistry curing.
  • Improves mechanical strength and thermal stability of polymer networks.
  • Liquid, colorless to light-yellow appearance.
  • Soluble in DMSO; may require sonication.
  • Typical purity around 95%.
  • Store refrigerated under inert atmosphere.