Learn More
Medchemexpress LLC Pentaerythritol tetra(3-mercaptopropionate) | 7575-23-7 | MFCD00022104 | 95.0% | 488.7 g/mol | C17H28O8S4 | 1 KG
SDP
Catalog No. 5000281232
Encompass_Preferred
Pentaerythritol tetra(3-mercaptopropionate) is a tetrafunctional mercapto-ester used as a crosslinker, stabilizer, and intermediate in specialty chemical and polymer applications. With four reactive thiol groups it facilitates rapid network formation and polymer modification in formulations and synthesis workflows. Typical applications include thiol-ene chemistry, resin formulation, and production of multifunctional materials.
  • Tetrafunctional thiol functionality enables efficient crosslinking.
  • Suitable for thiol-ene click reactions and polymer network formation.
  • Useful as an intermediate in specialty chemical synthesis.
  • Supplied at 95.0% purity for consistent performance.
  • Provides predictable reactivity for formulation and materials development.
Catalog No. 50-002-81232 Supplier Medchemexpress LLC Supplier No. HYW2503091KG
Pentaerythritol tetra(3-mercaptopropionate) is a tetrafunctional mercapto-ester used as a crosslinker, stabilizer, and intermediate in specialty chemical and polymer applications. With four reactive thiol groups it facilitates rapid network formation and polymer modification in formulations and synthesis workflows. Typical applications include thiol-ene chemistry, resin formulation, and production of multifunctional materials.
  • Tetrafunctional thiol functionality enables efficient crosslinking.
  • Suitable for thiol-ene click reactions and polymer network formation.
  • Useful as an intermediate in specialty chemical synthesis.
  • Supplied at 95.0% purity for consistent performance.
  • Provides predictable reactivity for formulation and materials development.