Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentaerythritol tetrabromide

Catalog No. P480125GA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P480125GA 25 g
P4801100GA 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P480125GA Supplier Sigma Aldrich Supplier No. P480125GA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentaerythritol tetrabromide

Specifications

CAS 3229-00-3
Melting Point 158°C to 160°C (lit.)
Density 2.596 g/mL (at 25°C (literature))
Boiling Point 305°C to 306°C (lit.)
Molecular Formula C5H8Br4
Linear Formula C(CH2Br)4
MDL Number MFCD00000210
Quantity 25 g
Synonym Tetrakis(bromomethyl)methane
Molecular Weight (g/mol) 387.73
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.