Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaerythritol Tetrakis(mercaptoacetate) 95.0+%, TCI America™
SDP

Catalog No. P0888500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0888500G 500 g
P088825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0888500G Supplier TCI America Supplier No. P0888500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10193-99-4 | C13H20O8S4 | 432.535 g/mol

Chemical Identifiers

CAS 10193-99-4
Molecular Formula C13H20O8S4
Molecular Weight (g/mol) 432.535
MDL Number MFCD00022080
InChI Key RUDUCNPHDIMQCY-UHFFFAOYSA-N
Synonym Pentaerythritol Tetrathioglycolate
PubChem CID 82441
IUPAC Name [3-(2-sulfanylacetyl)oxy-2,2-bis[(2-sulfanylacetyl)oxymethyl]propyl] 2-sulfanylacetate
SMILES C(C(=O)OCC(COC(=O)CS)(COC(=O)CS)COC(=O)CS)S

Specifications

Color Colorless
Boiling Point 250°C
Quantity 500 g
Formula Weight 432.54
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Pentaerythritol Tetrakis(mercaptoacetate)
EINECSNumber (2)-3334
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.