Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaerythritol Tetrastearate, MP Biomedicals

Catalog No. ICN217303 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217303 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217303 Supplier MP Biomedicals, Inc Supplier No. 0521730301

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 115-83-3
Molecular Formula C77H148O8
Molecular Weight (g/mol) 1202.02
MDL Number MFCD00041922
InChI Key OCKWAZCWKSMKNC-UHFFFAOYSA-N
Synonym pentaerythrityl tetrastearate, pentaerythritol tetrastearate, unii-w9q3dzs0eg, stearic acid, neopentanetetrayl ester, w9q3dzs0eg, 3-octadecanoyloxy-2,2-bis octadecanoyloxymethyl propyl octadecanoate, octadecanoic acid, 2,2-bis 1-oxooctadecyl oxy methyl-1,3-propanediyl ester, pe tetrastearate, pentaerythrittetrastearat
PubChem CID 61037
IUPAC Name 3-(octadecanoyloxy)-2,2-bis[(octadecanoyloxy)methyl]propyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)(COC(=O)CCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCC

Specifications

Quantity 1 kg
Formula Weight 1202.02
Chemical Name or Material Pentaerythritol Tetrastearate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.