Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentaethylene glycol | 4792-15-8 | 238.28 | 25 G
SDP

Catalog No. 5000449187
Encompass_Preferred

Pentaethylene glycol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to specifically degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.

  • Purity: 99.58%
  • Formula: C10H22O6
  • Appearance: Liquid (density: 1.126 g/cm3), colorless to light yellow
  • Cas no.: 4792-15-8
  • Application: Used in the synthesis of PROTACs

Catalog No. 50-004-49187 Supplier Medchemexpress LLC Supplier No. 25GHYW004447
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentaethylene glycol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to specifically degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.

  • Purity: 99.58%
  • Formula: C10H22O6
  • Appearance: Liquid (density: 1.126 g/cm3), colorless to light yellow
  • Cas no.: 4792-15-8
  • Application: Used in the synthesis of PROTACs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.