Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentaethylene glycol di(p-toluenesulfonate)

Catalog No. 3095835G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-958-35G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-958-35G Supplier Sigma Aldrich Supplier No. 3095835G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentaethylene glycol di(p-toluenesulfonate)

Specifications

CAS 41024-91-3
Density 1.26 g/mL (at 25°C (literature))
Molecular Formula C24H34O10S2
Refractive Index n20/D 1.524 (literature)
Linear Formula CH3C6H4SO3(CH2CH2O)5 SO2C6H4CH3
MDL Number MFCD00012204
Quantity 100 mg
Synonym 3,6,9,12-Tetraoxatetradecane-1,14-diyl ditosylate; Pentaethylene glycol ditosylate
Molecular Weight (g/mol) 546.65
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.