Learn More
Medchemexpress LLC Pentaethylene glycol di(p-toluenesulfonate) | 41024-91-3 | 546.65 | 10 G
SDP
Catalog No. 5000445399
Encompass_Preferred
Pentaethylene glycol di(p-toluenesulfonate) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-004-45399 Supplier Medchemexpress LLC Supplier No. HYW00444710G
Pentaethylene glycol di(p-toluenesulfonate) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only