Learn More
Medchemexpress LLC Pentaethylene glycol di(p-toluenesulfonate) | 41024-91-3 | 546.65 | 5 G
SDP
Catalog No. 5000448821
Encompass_Preferred
Pentaethylene glycol di(p-toluenesulfonate) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system, selectively degrading target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein via a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-48821 Supplier Medchemexpress LLC Supplier No. HYW0044475G
Pentaethylene glycol di(p-toluenesulfonate) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system, selectively degrading target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein via a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins