Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentaethylene glycol monomethyl ether | 23778-52-1 | MFCD00045995 | 252.30 g/mol | C11H24O6 | 5 G
SDP

Catalog No. 5000210171
Encompass_Preferred

Pentaethylene glycol monomethyl ether is a polyethylene glycol (PEG) derivative used as a PEG-based PROTAC linker in research and small-scale synthesis. It is a colorless, low-volatility liquid commonly used to introduce a methoxy-terminated PEG5 spacer in conjugation and linker chemistry.

  • PEG-based linker suitable for PROTAC synthesis and bioconjugation
  • Molecular formula C11H24O6, molecular weight 252.30 g/mol
  • Available in small laboratory pack sizes, for example 5 G
  • Soluble in common organic solvents and used as a spacer and solubilizing unit

Catalog No. 50-002-10171 Supplier Medchemexpress LLC Supplier No. HYW0427135G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentaethylene glycol monomethyl ether is a polyethylene glycol (PEG) derivative used as a PEG-based PROTAC linker in research and small-scale synthesis. It is a colorless, low-volatility liquid commonly used to introduce a methoxy-terminated PEG5 spacer in conjugation and linker chemistry.

  • PEG-based linker suitable for PROTAC synthesis and bioconjugation
  • Molecular formula C11H24O6, molecular weight 252.30 g/mol
  • Available in small laboratory pack sizes, for example 5 G
  • Soluble in common organic solvents and used as a spacer and solubilizing unit

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.