Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentaethylene glycol monomethyl ether | 23778-52-1 | MFCD00045995 | 99.8% | 252.30 | C11H24O6 | 25 G
SDP

Catalog No. 5000208830
Encompass_Preferred

Pentaethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based monomethyl ether used as a short, flexible linker and spacer in medicinal chemistry and PROTAC synthesis. It is a colorless to light-yellow liquid (C11H24O6, MW 252.30) supplied in laboratory pack sizes for synthetic applications.

  • Peg-based monomethyl ether suitable for linker design in PROTACs.
  • High purity for synthetic and conjugation reactions.
  • Available in small lab pack sizes (5 g, 25 g, 100 g).
  • Storage recommendations provided for pure material and solutions to maintain stability.
  • Soluble in common organic solvents such as DMSO for use in reaction setups.

Catalog No. 50-002-08830 Supplier Medchemexpress LLC Supplier No. 25GHYW042713
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentaethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based monomethyl ether used as a short, flexible linker and spacer in medicinal chemistry and PROTAC synthesis. It is a colorless to light-yellow liquid (C11H24O6, MW 252.30) supplied in laboratory pack sizes for synthetic applications.

  • Peg-based monomethyl ether suitable for linker design in PROTACs.
  • High purity for synthetic and conjugation reactions.
  • Available in small lab pack sizes (5 g, 25 g, 100 g).
  • Storage recommendations provided for pure material and solutions to maintain stability.
  • Soluble in common organic solvents such as DMSO for use in reaction setups.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.