Learn More
Medchemexpress LLC Pentaethylene glycol monomethyl ether | 23778-52-1 | MFCD00045995 | 99.9% | 252.30 g/mol | C11H24O6 | 100 G
SDP
Catalog No. 5000210083
Encompass_Preferred
Pentaethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based linker intended for research use in proteolysis targeting chimera (PROTAC) synthesis. It serves as a flexible spacer between ligands to enable recruitment of E3 ubiquitin ligases for targeted protein degradation.
  • Peg-based PROTAC linker suitable for connecting ligands in bifunctional degraders.
  • High purity, supporting reliable synthetic workflows.
  • Good solubility in DMSO (200 mg/mL) with ultrasonic assistance if required.
  • Available in multiple pack sizes for research-scale needs (5 g-100 g, bulk).
  • Storage stability in pure form at -20°C (up to 3 years) for long-term use.
Catalog No. 50-002-10083 Supplier Medchemexpress LLC Supplier No. HYW042713100G
Pentaethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based linker intended for research use in proteolysis targeting chimera (PROTAC) synthesis. It serves as a flexible spacer between ligands to enable recruitment of E3 ubiquitin ligases for targeted protein degradation.
  • Peg-based PROTAC linker suitable for connecting ligands in bifunctional degraders.
  • High purity, supporting reliable synthetic workflows.
  • Good solubility in DMSO (200 mg/mL) with ultrasonic assistance if required.
  • Available in multiple pack sizes for research-scale needs (5 g-100 g, bulk).
  • Storage stability in pure form at -20°C (up to 3 years) for long-term use.