Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoroanilinium Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. P16265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16265G 5 g
P16261G 1 g
P162625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P16265G Supplier TCI America Supplier No. P16265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 912823-79-1
Molecular Formula C7H3F8NO3S
Synonym Pentafluoroanilinium Triflate, (Pentafluorophenyl)ammonium Trifluoromethanesulfonate, PFPAT

Specifications

Quantity 5 g
UN Number 1759
Formula Weight 333.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluoroanilinium Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.