Learn More
Strem, An Ascensus Company CAS# 653-37-2. 5g. Pentafluorobenzaldehyde, 98%. MFCD00003303
SDP
Catalog No. 5090109360
Encompass
CAS# 653-37-2. 5g. Pentafluorobenzaldehyde, 98%. MFCD00003303. Molecular Weight: 196.07. Molecular Formula: C6F5CHO. Color/form: colorless to pale yellow solid Strem# 09-5440. http://www.strem.com/catalog/v/09-5440/
Catalog No. 50-901-09360 Supplier Strem, An Ascensus Company Supplier No. 0954405G
CAS# 653-37-2. 5g. Pentafluorobenzaldehyde, 98%. MFCD00003303. Molecular Weight: 196.07. Molecular Formula: C6F5CHO. Color/form: colorless to pale yellow solid Strem# 09-5440. http://www.strem.com/catalog/v/09-5440/