Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzaldehyde, 98%

Catalog No. AAA1549006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1549006 5 g
AAA1549014 25 g
AAA1549022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1549006 Supplier Thermo Scientific Chemicals Supplier No. A1549006
  • $79.20
    $63.65 / Each of 1

    Save $15.55

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 653-37-2 | C7HF5O | 196.076 g/mol

Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.

Solubility
Sparingly Soluble in water (0.027 g/L at 25°C).

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, air, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-37-2
Molecular Formula C7HF5O
Molecular Weight (g/mol) 196.076
MDL Number MFCD00003303
InChI Key QJXCFMJTJYCLFG-UHFFFAOYSA-N
Synonym pentafluorobenzaldehyde, perfluorobenzaldehyde, benzaldehyde, pentafluoro, benzaldehyde, 2,3,4,5,6-pentafluoro, unii-s9yie3jz4k, pentafluor-benzaldehyd, s9yie3jz4k, pubchem4253, pentafluoro-benzaldehyde, acmc-209nqh
PubChem CID 69558
IUPAC Name 2,3,4,5,6-pentafluorobenzaldehyde
SMILES C(=O)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point 24°C to 27°C
Density 1.63
Boiling Point 164°C to 166°C
Flash Point 77°C (170°F)
Refractive Index 1.451
Quantity 5 g
UN Number UN2811
Beilstein 1876550
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble in water (0.027g/L at 25°C).
Formula Weight 196.07
Percent Purity ≥98%
Chemical Name or Material Pentafluorobenzaldehyde
Show More Show Less
Hazard Category H227-H315-H319-H330-H335
Hazard Statement GHS H Statement
H331-H315-H319-H335
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 211-502-6
RTECSNumber CU7542000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.