Learn More
Pentafluorobenzaldehyde, 98%
Description
Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.
Solubility
Sparingly Soluble in water (0.027 g/L at 25°C).
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, air, bases.
Specifications
Specifications
| Melting Point | 24°C to 27°C |
| Density | 1.63 |
| Boiling Point | 164°C to 166°C |
| Flash Point | 77°C (170°F) |
| Refractive Index | 1.451 |
| Quantity | 25 g |
| UN Number | UN2811 |
| Beilstein | 1876550 |
| Sensitivity | Air Sensitive |
| Solubility Information | Sparingly Soluble in water (0.027g/L at 25°C). |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319-H330-H335 |
| Hazard Statement | GHS H Statement H331-H315-H319-H335 Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P235-P260-P264b-P271-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 211-502-6 |
| RTECSNumber | CU7542000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.