Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentafluorobenzenesulfonyl chloride

Catalog No. 1037645G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-376-45G 5 g
10-376-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-376-45G Supplier Sigma Aldrich Supplier No. 1037645G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Pentafluorobenzenesulfonyl chloride reacts with benzene and thiophene derivatives in the presence of a ruthenium (ii) catalyst to form corresponding perfluorophenylated compounds.pentafluorobenzenesulfonyl chloride was used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an o-dealkylated metabolite of fluoxetine in human liver microsomes
  • Pentafluorobenzenesulfonyl chloride was used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection.pentafluorobenzenesulfonyl chloride is derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detectio

Specifications

CAS 832-53-1
Density 1.796 g/mL (at 25°C (literature))
Boiling Point 210°C to 211°C (lit.)
Molecular Formula C6ClF5O2S
Refractive Index n20/D 1.479 (literature)
Linear Formula C6F5SO2Cl
MDL Number MFCD00007427
Quantity 5 g
Molecular Weight (g/mol) 266.57
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.