Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzenesulfonyl chloride, 99%

Catalog No. AC147110050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147110050 5 g Glass bottle
AC147110250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC147110050 Supplier Thermo Scientific Chemicals Supplier No. 147110050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 832-53-1 | C6ClF5O2S | 266.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 832-53-1
Molecular Formula C6ClF5O2S
Molecular Weight (g/mol) 266.57
MDL Number MFCD00007427
InChI Key UOJCTEGNHXRPKO-UHFFFAOYSA-N
Synonym pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
PubChem CID 70026
IUPAC Name 2,3,4,5,6-pentafluorobenzenesulfonyl chloride
SMILES C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F

Specifications

Density 1.7960g/mL
Boiling Point 211°C
Flash Point 90°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4780 to 1.48
Linear Formula C6F5SO2Cl
Quantity 5 g
Specific Gravity 1.796
Formula Weight 266.57
Percent Purity 99%
Chemical Name or Material Pentafluorobenzenesulfonyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 212-620-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.