Learn More
Strem, An Ascensus Company CAS# 602-94-8. 1g. Pentafluorobenzoic acid, 99%. MFCD00002406
SDP
Catalog No. 5090109364
Encompass
CAS# 602-94-8. 1g. Pentafluorobenzoic acid, 99%. MFCD00002406. Molecular Weight: 212.07. Molecular Formula: C6F5COOH. Color/form: white xtl. Strem# 09-5500. http://www.strem.com/catalog/v/09-5500/
Catalog No. 50-901-09364 Supplier Strem, An Ascensus Company Supplier No. 0955001G
Add to Cart
Add to Cart
CAS# 602-94-8. 1g. Pentafluorobenzoic acid, 99%. MFCD00002406. Molecular Weight: 212.07. Molecular Formula: C6F5COOH. Color/form: white xtl. Strem# 09-5500. http://www.strem.com/catalog/v/09-5500/