Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoronitrobenzene, MP Biomedicals

Catalog No. ICN226004 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN226004 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN226004 Supplier MP Biomedicals, Inc Supplier No. 0522600401

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 880-78-4
Molecular Formula C6F5NO2
Molecular Weight (g/mol) 213.063
InChI Key INUOFQAJCYUOJR-UHFFFAOYSA-N
Synonym pentafluoronitrobenzene, nitropentafluorobenzene, 2,3,4,5,6-pentafluoronitrobenzene, benzene, pentafluoronitro, 1,2,3,4,5-pentafluoro-6-nitro-benzene, 2,3,4,5,6-pentafluoro-1-nitrobenzene, pubchem1060, acmc-1bm0y
PubChem CID 70154
IUPAC Name 1,2,3,4,5-pentafluoro-6-nitrobenzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 159
Chemical Name or Material Pentafluoronitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.