Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoronitrobenzene 97.0+%, TCI America™
SDP

Catalog No. P12285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12285G 5 g
P122825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12285G Supplier TCI America Supplier No. P12285G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 880-78-4
Molecular Formula C6F5NO2
Molecular Weight (g/mol) 213.063
MDL Number MFCD00007049
InChI Key INUOFQAJCYUOJR-UHFFFAOYSA-N
Synonym pentafluoronitrobenzene, nitropentafluorobenzene, 2,3,4,5,6-pentafluoronitrobenzene, benzene, pentafluoronitro, 1,2,3,4,5-pentafluoro-6-nitro-benzene, 2,3,4,5,6-pentafluoro-1-nitrobenzene, pubchem1060, acmc-1bm0y
PubChem CID 70154
IUPAC Name 1,2,3,4,5-pentafluoro-6-nitrobenzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 160°C
Quantity 5 g
Formula Weight 213.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluoronitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.