Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenol, 99%

Catalog No. AAA1557406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1557406 5 g
AAA1557414 25 g
AAA1557422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1557406 Supplier Thermo Scientific Chemicals Supplier No. A1557406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 771-61-9 | C6HF5O | 184.07 g/mol

Pentafluorophenol is used in the preparation of pentafluorophenyl esters for peptide synthesis and vulcanized polymers. It is also used for peptide coupling and heterocyclic acid derivatives. It is involved in the preparation of pentafluorophenyl formate which acts as a formulating agent for amines and amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorophenol is used in the preparation of pentafluorophenyl esters for peptide synthesis and vulcanized polymers. It is also used for peptide coupling and heterocyclic acid derivatives. It is involved in the preparation of pentafluorophenyl formate which acts as a formulating agent for amines and amino acids.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, bases, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-61-9
Molecular Formula C6HF5O
Molecular Weight (g/mol) 184.07
MDL Number MFCD00002156
InChI Key XBNGYFFABRKICK-UHFFFAOYSA-N
Synonym pentafluorophenol, phenol, pentafluoro, perfluorophenol, unii-a2ycf0yuha, a2ycf0yuha, 2,3,4,5,6-pentafluorohydroxybenzene, c6f5oh, 2,3,4,5,6-pentafluoro-phenol, phenol, 2,3,4,5,6-pentafluoro, hopfp
PubChem CID 13041
IUPAC Name 2,3,4,5,6-pentafluorophenol
SMILES OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 32°C to 36°C
Density 1.757
Boiling Point 143°C
Flash Point 72°C (161°F)
Refractive Index 1.427
Quantity 5 g
Beilstein 1912584
Solubility Information Soluble in water.
Formula Weight 184.06
Percent Purity 99%
Chemical Name or Material Pentafluorophenol
Show More Show Less
Hazard Category H302+H312+H332-H314-H318-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 212-235-8
RTECSNumber SM6680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.