Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenyl 2-[4-(tert-butoxycarbonyl)piperazin-1-yl]isonicotinate, 90%, Thermo Scientific™

Catalog No. CC50626DA
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC50626DA 1 g Amber Glass Bottle
CC50626DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC50626DA Supplier Thermo Scientific Chemicals Supplier No. CC50626DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 944450-81-1
Molecular Formula C21H20F5N3O4
Molecular Weight (g/mol) 473.4
MDL Number MFCD09966147
InChI Key XHATYMDMSNXMKQ-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-4-tert-butoxycarbonyl piperazin-1-yl isonicotinate, tert-butyl 4-4-pentafluorophenoxycarbonyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-pentafluorophenoxy carbonyl pyridin-2-yl piperazine-1-carboxylate, 4-4-oxo-2,3,4,5,6-pentafluorophenoxy methyl-2-pyridinyl-1-piperazinecarboxylic acid tert-butyl ester, tert-butyl 4-4-2,3,4,5,6-pentafluorophenoxycarbonyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-2,3,4,5,6-pentakis fluoranyl phenoxy carbonylpyridin-2-yl piperazine-1-carboxylate
PubChem CID 43811038
IUPAC Name tert-butyl 4-[4-(2,3,4,5,6-pentafluorophenoxy)carbonylpyridin-2-yl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=CC(=C2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 473.4
Percent Purity 90%
Chemical Name or Material Pentafluorophenyl 2-[4-(tert-butoxycarbonyl)piperazin-1-yl]isonicotinate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.