Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenyl 2-pyrid-3-yl-1,3-thiazole-4-carboxylate, 97%, Thermo Scientific™

Catalog No. CC13726DA
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC13726DA 1 g
CC13726CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC13726DA Supplier Thermo Scientific Chemicals Supplier No. CC13726DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 906352-58-7
Molecular Formula C15H5F5N2O2S
Molecular Weight (g/mol) 372.27
MDL Number MFCD09025821
InChI Key LHRLGPDYSCUADI-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-pyrid-3-yl-1,3-thiazole-4-carboxylate, pentafluorophenyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate, 2,3,4,5,6-pentafluorophenyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate, perfluorophenyl 2-pyridin-3-yl thiazole-4-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate, 2-3-pyridinyl-4-thiazolecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 18525705
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CSC(=N2)C2=CN=CC=C2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2-pyridin-3-yl-1,3-thiazole-4-carboxylate
Formula Weight 372.27
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 2-pyrid-3-yl-1, 3-thiazole-4-carboxylate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.