Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentafluorophenyl acrylate

Catalog No. 7530921G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-309-21G 1 g
75-309-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-309-21G Supplier Sigma Aldrich Supplier No. 7530921G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentafluorophenyl acrylate may be used in the preparation of highly porous polymers (poly HIPs). Pentafluorophenyl acrylate (PFPA) can undergo reversible addition fragmentation transfer (RAFT) polymerization with oligoethylene glycol acrylate (OEGA) or diethylene glycol acrylate (DEGA).

Specifications

CAS 71195-85-2
Density 1.446
Molecular Formula C9H3F5O2
Refractive Index n/D 1.434
Linear Formula C9H3F5O2
Quantity 1 g
MDL Number MFCD00042330
Synonym 2-Propenoic acid pentafluorophenyl ester
Molecular Weight (g/mol) 238.11
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.