Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Pentafluorophenyl)diphenylphosphine 93.0+%, TCI America™
SDP

Catalog No. P11765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11765G 5 g
P11761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11765G Supplier TCI America Supplier No. P11765G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5525-95-1
Molecular Formula C18H10F5P
Molecular Weight (g/mol) 352.24
MDL Number MFCD00000290
InChI Key KUTXTUCJQJPJBH-UHFFFAOYSA-N
Synonym diphenyl pentafluorophenyl phosphine, pentafluorophenyl diphenylphosphine, perfluorophenyl diphenylphosphine, phosphine, pentafluorophenyl diphenyl, pentafluorophenyl diphenylphosphane, pubchem6416, acmc-1aogr, pentafluorodiphenylphosphine, diphenylpentafluorophenyphosphine, pentafluorophenyldiphenylphosphine
PubChem CID 300563
IUPAC Name (2,3,4,5,6-pentafluorophenyl)diphenylphosphane
SMILES FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=CC=CC=C2)C(F)=C1F

Specifications

Melting Point 70°C
Color White
Quantity 5 g
Formula Weight 352.24
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Pentafluorophenyl)diphenylphosphine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.